Gene Transcripts

Gene Symbol
Aven
Entrez Gene ID
74268
Gene Name
apoptosis, caspase activation inhibitor
Gene Aliases
1700013A01Rik, 1700056A21Rik, mAven-L, mAven-S
Chromosome Location
Chr. 2: 112492964 - 112631253 on Build GRCm38
UniGene ID
Mm.315125
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028844.3 NP_083120.2 2 380
GenBank mRNA AK006808.1 - 2 381
BC100725.1 - 3 243 40045263
BC100735.1 - 3 257

View Full Product Details