Gene Transcripts

Gene Symbol
Tcam1
Entrez Gene ID
75870
Gene Name
testicular cell adhesion molecule 1
Gene Aliases
4930570F09Rik, AI429105
Chromosome Location
Chr. 11: 106273079 - 106288158 on Build GRCm38
UniGene ID
Mm.159608
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029467.3 NP_083743.2 3 298
GenBank mRNA AK016263.1 - 3 168
AK076581.1 - 3 218
BC050746.1 - 222 6741794

View Full Product Details