Gene Transcripts

Gene Symbol
Mpst
Entrez Gene ID
246221
Gene Name
mercaptopyruvate sulfurtransferase
Gene Aliases
Mst
Chromosome Location
Chr. 15: 78406712 - 78414015 on Build GRCm38
UniGene ID
Mm.294215
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162492.1 NP_001155964.1 3 800
NM_001162493.1 NP_001155965.1 3 770
NM_138670.3 NP_619611.3 3 904
GenBank mRNA AK136571.1 - 2 799
AY974575.1 - 1 66
BC004079.1 - 2 745 3592659
BC006066.1 - 2 745
BC094469.1 - 3 904 5009481

View Full Product Details