Gene Transcripts

Gene Symbol
Sec16a
Entrez Gene ID
227648
Gene Name
SEC16 homolog A (S. cerevisiae)
Gene Aliases
AU024582, C230052J16Rik
Chromosome Location
Chr. 2: 26409431 - 26446162 on Build GRCm38
UniGene ID
Mm.206371
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153125.2 NP_694765.2 16 5609
GenBank mRNA AK122242.2 - 9 1046
BC034649.1 - 6 667
BC042603.1 - 10 1125
BC059898.1 - 7 1212

View Full Product Details