Gene Transcripts

Gene Symbol
Amot
Entrez Gene ID
27494
Gene Name
angiomotin
Gene Aliases
CAG-2, D0Kist1, Sii6
Chromosome Location
Chr. X: 145446424 - 145505541 on Build GRCm38
UniGene ID
Mm.100068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290274.1 NP_001277203.1 1 789
NM_153319.3 NP_695231.3 1 789
GenBank mRNA AF461135.1 - 1 196
BC057638.1 - 1 244 2631191

View Full Product Details