Gene Transcripts
- Gene Symbol
- Zdhhc17
- Entrez Gene ID
- 320150
- Gene Name
- zinc finger, DHHC domain containing 17
- Gene Aliases
- A230053P19Rik, BB187739, D130071N24Rik, Hip14
- Chromosome Location
- Chr. 10: 110941779 - 111010066 on Build GRCm38
- UniGene ID
- Mm.339281
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172554.2 |
NP_766142.2
|
2, 3 |
233 |
|
GenBank mRNA |
AK038670.1 |
- |
2, 3 |
177 |
|
AK038681.1 |
- |
2, 3 |
177 |
|
AK039575.1 |
- |
2, 3 |
234 |
|
AK046891.1 |
- |
2, 3 |
175 |
|
AK122406.1 |
- |
2, 3 |
307 |
|
AK134967.1 |
- |
2, 3 |
182 |
|
AK162922.1 |
- |
2, 3 |
171 |
|
BC027101.1 |
- |
2, 3 |
162 |
|
BC058772.1 |
- |
2, 3 |
174 |
|
BC080664.1 |
- |
2, 3 |
172 |
|
BC117788.1 |
- |
2, 3 |
175 |
40092137 |
BC117789.2 |
- |
2, 3 |
175 |
40092140 |
View Full Product Details