Gene Transcripts
- Gene Symbol
- IRAK4
- Entrez Gene ID
- 51135
- Gene Name
- interleukin 1 receptor associated kinase 4
- Gene Aliases
- IPD1, IRAK-4, NY-REN-64, REN64
- Chromosome Location
- Chr. 12: 43758913 - 43789543 on Build GRCh38
- UniGene ID
- Hs.138499
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114182.2 |
NP_001107654.1
|
3, 4 |
299 |
|
NM_016123.3 |
NP_057207.2
|
2, 3 |
251 |
|
XM_005268943.3 |
XP_005269000.1
|
2, 3 |
356 |
|
XM_005268944.4 |
XP_005269001.1
|
2, 3 |
320 |
|
XM_005268945.4 |
XP_005269002.1
|
2, 3 |
564 |
|
XM_006719438.3 |
XP_006719501.1
|
3, 4 |
612 |
|
XM_006719440.2 |
XP_006719503.1
|
2, 3 |
214 |
|
XM_011538431.2 |
XP_011536733.1
|
3, 4 |
368 |
|
XM_011538432.1 |
XP_011536734.1
|
3, 4 |
404 |
|
XM_011538433.2 |
XP_011536735.1
|
2, 3 |
983 |
|
XM_017019390.1 |
XP_016874879.1
|
4, 5 |
485 |
|
GenBank mRNA |
AF155118.1 |
- |
2, 3 |
218 |
|
AF445802.1 |
- |
1, 2 |
169 |
|
AK000528.1 |
- |
2, 3 |
198 |
|
AK314288.1 |
- |
2, 3 |
240 |
|
AY283670.1 |
- |
1, 2 |
169 |
|
AY283671.1 |
- |
1, 2 |
169 |
|
AY340962.1 |
- |
2, 3 |
234 |
|
AY340963.1 |
- |
3, 4 |
282 |
|
AY730682.1 |
- |
1, 2 |
169 |
|
BC013316.1 |
- |
2, 3 |
239 |
4287014 |
KU178631.1 |
- |
1, 2 |
169 |
|
View Full Product Details