Gene Transcripts
- Gene Symbol
- SENP3-EIF4A1
- Entrez Gene ID
- 100533955
- Gene Name
- SENP3-EIF4A1 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 17: 7561992 - 7579006 on Build GRCh38
- UniGene ID
- Hs.129673
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037926.1 |
|
17 |
3149 |
|
- Gene Symbol
- EIF4A1
- Entrez Gene ID
- 1973
- Gene Name
- eukaryotic translation initiation factor 4A1
- Gene Aliases
- DDX2A, EIF-4A, EIF4A, eIF-4A-I, eIF4A-I
- Chromosome Location
- Chr. 17: 7572706 - 7579006 on Build GRCh38
- UniGene ID
- Hs.129673
- Species
- Homo sapiens
View Full Product Details