Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QSOX1
Entrez Gene ID
838067
Gene Name
quiescin-sulfhydryl oxidase 1
Gene Aliases
AT1G15020, ATQSOX1, QSO2, T15D22.7, T15D22_7, quiescin-sulfhydryl oxidase 1
Chromosome Location
Chr.1: 5172232 - 5176479 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332143.1 NP_001321934.1 5 - 6 899 100
NM_101371.5 NP_172955.1 5 - 6 949 100
NM_179333.2 NP_849664.1 5 - 6 899 100
GenBank mRNA AY062528.1 - 5 - 6 690 100
BT001218.1 - 5 - 6 575 100

View Full Product Details