Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- WAKL7
- Entrez Gene ID
-
838179
- Gene Name
- wall associated kinase-like 7
- Gene Aliases
-
AT1G16090, T24D18.19, T24D18_19, wall associated kinase-like 7
- Chromosome Location
-
Chr.1: 5516259 - 5517822 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_101477.2 |
NP_173061.2 |
2 - 3 |
767 |
130 |
View Full Product Details