Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATB BETA
Entrez Gene ID
838348
Gene Name
Protein phosphatase 2A, regulatory subunit PR55
Gene Aliases
AT1G17720, F11A6.6, F11A6_6
Chromosome Location
Chr.1: 6093185 - 6098443 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101634.4 NP_564033.1 13 - 14 1739 87
NM_179350.4 NP_849681.1 13 - 14 1740 87
GenBank mRNA AF370519.1 - 13 - 14 1630 87
AK226540.1 - 13 - 14 1683 87
AY045665.1 - 13 - 14 1630 87
BT000645.1 - 13 - 14 1365 87
BT002183.1 - 13 - 14 1362 87
U40161.1 - 13 - 14 1451 87

View Full Product Details