Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATB BETA
Entrez Gene ID
838348
Gene Name
Protein phosphatase 2A, regulatory subunit PR55
Gene Aliases
AT1G17720, F11A6.6, F11A6_6
Chromosome Location
Chr.1: 6093185 - 6098443 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101634.4 NP_564033.1 7 - 8 1181 130
NM_179350.4 NP_849681.1 7 - 8 1182 130
GenBank mRNA AF370519.1 - 7 - 8 1072 130
AK226540.1 - 7 - 8 1125 130
AY045665.1 - 7 - 8 1072 130
BT000645.1 - 7 - 8 807 130
BT002183.1 - 7 - 8 804 130
U40161.1 - 7 - 8 893 130

View Full Product Details