Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC1
Entrez Gene ID
838393
Gene Name
laccase 1
Gene Aliases
AT1G18140, ATLAC1, T10F20.14, laccase 1
Chromosome Location
Chr.1: 6238839 - 6241510 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101674.4 NP_173252.2 2 - 3 371 92
GenBank mRNA AK227320.1 - 2 - 3 297 92

View Full Product Details