Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATGRIP
Entrez Gene ID
836733
Gene Name
Golgi-localized GRIP domain-containing protein
Gene Aliases
AT5G66030, GRIP, K2A18.10, K2A18_10
Chromosome Location
Chr.5: 26404893 - 26410355 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345737.1 NP_001332639.1 15 - 16 2147 87
NM_001345738.1 NP_001332640.1 15 - 16 2122 87
NM_126000.2 NP_569025.2 14 - 15 1852 87
NM_180946.3 NP_851277.1 15 - 16 2182 87

View Full Product Details