Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK28
Entrez Gene ID
836753
Gene Name
calcium-dependent protein kinase 28
Gene Aliases
AT5G66210, K2A18.29, K2A18_29, calcium-dependent protein kinase 28
Chromosome Location
Chr.5: 26456142 - 26459654 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085337.1 NP_001078806.1 6 - 7 1018 88
NM_001126036.2 NP_001119508.1 6 - 7 1018 88
NM_001345751.1 NP_001331199.1 6 - 7 1018 88
NM_001345752.1 NP_001331200.1 6 - 7 1048 88
NM_126019.4 NP_201422.1 7 - 8 910 88
NM_180949.3 NP_851280.1 6 - 7 1018 88
GenBank mRNA AK227052.1 - 6 - 7 843 88
AK317109.1 - 6 - 7 837 88
AY139991.1 - 6 - 7 842 88
BT010380.1 - 6 - 7 828 88

View Full Product Details