Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAR5
Entrez Gene ID
836795
Gene Name
DA1-related protein 5
Gene Aliases
AT5G66630, DA1-related protein 5, K1F13.31, K1F13_31
Chromosome Location
Chr.5: 26592648 - 26595922 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126061.3 NP_201464.2 7 - 8 1682 73

View Full Product Details