Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-B4
Entrez Gene ID
814759
Gene Name
phloem protein 2-B4
Gene Aliases
AT2G02280, AtPP2-B4, T16F16.7, T16F16_7, phloem protein 2-B4
Chromosome Location
Chr.2: 600491 - 601439 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126286.1 NP_178334.1 1 - 2 204 101

View Full Product Details