Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-B6
Entrez Gene ID
814762
Gene Name
phloem protein 2-B6
Gene Aliases
AT2G02310, AtPP2-B6, T16F16.10, T16F16_10, phloem protein 2-B6
Chromosome Location
Chr.2: 608586 - 610283 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126289.2 NP_178337.1 2 - 3 643 89

View Full Product Details