Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRS1
Entrez Gene ID
819671
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT3G05090, LATERAL ROOT STIMULATOR 1, T12H1.5, T12H1_5
Chromosome Location
Chr.3: 1417893 - 1423673 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337562.1 NP_001325515.1 15 - 16 2175 70
NM_111381.3 NP_566246.1 15 - 16 2292 70
NM_180185.3 NP_850516.1 15 - 16 2290 70
GenBank mRNA AK226843.1 - 15 - 16 2250 70
AY056292.1 - 15 - 16 2200 70
AY127022.1 - 15 - 16 2243 70
AY142674.1 - 14 - 15 1891 70
BT000631.1 - 14 - 15 1891 70

View Full Product Details