Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATG10
Entrez Gene ID
819941
Gene Name
autophagocytosis-associated family protein
Gene Aliases
AT3G07525, ATATG10, autophagy 10
Chromosome Location
Chr.3: 2398951 - 2400603 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180201.3 NP_850532.1 2 - 3 439 128
NM_180202.1 NP_850533.1 2 - 3 241 128
GenBank mRNA AY065244.1 - 2 - 3 238 128
AY142587.1 - 2 - 3 216 128

View Full Product Details