Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KINESIN-13A
Entrez Gene ID
820915
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT3G16630, ATKINESIN-13A
Chromosome Location
Chr.3: 5662375 - 5667951 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112536.3 NP_188285.1 4 - 5 935 100
NM_180267.2 NP_850598.1 4 - 5 913 100
GenBank mRNA AK226569.1 - 4 - 5 790 100
AY056129.1 - 4 - 5 771 100

View Full Product Details