Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDH-E1 ALPHA
Entrez Gene ID
839429
Gene Name
pyruvate dehydrogenase E1 alpha
Gene Aliases
AT1G01090, T25K16.8, T25K16_8, pyruvate dehydrogenase E1 alpha
Chromosome Location
Chr.1: 47234 - 49304 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_099991.4 NP_171617.1 2 - 3 1150 64
GenBank mRNA AK226909.1 - 2 - 3 1129 64
AY052721.1 - 2 - 3 1127 64
AY063724.1 - 2 - 3 1012 64
U80185.1 - 2 - 3 1129 64

View Full Product Details