Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P58IPK
Entrez Gene ID
831917
Gene Name
DnaJ P58IPK-like protein
Gene Aliases
AT5G03160, ATP58IPK, F15A17.190, F15A17_190, homolog of mamallian P58IPK
Chromosome Location
Chr.5: 750101 - 752805 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120394.4 NP_195936.1 4 - 5 844 68
GenBank mRNA AY085329.1 - 4 - 5 797 68

View Full Product Details