Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP10
Entrez Gene ID
816569
Gene Name
nucleolar RNA-binding Nop10p family protein
Gene Aliases
AT2G20490, EDA27, EMBRYO SAC DEVELOPMENT ARREST 27, T13C7.8, T13C7_8
Chromosome Location
Chr.2: 8831681 - 8832929 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335686.1 NP_001325224.1 1 - 2 240 77
NM_127608.4 NP_565472.1 2 - 3 185 77
GenBank mRNA AY087879.1 - 2 - 3 144 77

View Full Product Details