Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAS1
Entrez Gene ID
824568
Gene Name
FKBP-type peptidyl-prolyl cis-trans isomerase family protein
Gene Aliases
AT3G54010, DEI1, PASTICCINO 1
Chromosome Location
Chr.3: 20000821 - 20005280 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115261.4 NP_566993.1 12 - 13 1312 119
NM_180370.3 NP_850701.1 11 - 12 1342 119
GenBank mRNA AY056265.1 - 12 - 13 1244 119
AY096635.1 - 12 - 13 1091 119

View Full Product Details