Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPB
Entrez Gene ID
831847
Gene Name
Transcription factor DP
Gene Aliases
AT5G03415, ATDPB, DPB TRANSCRIPTION FACTOR
Chromosome Location
Chr.5: 842289 - 845454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_180426.3 NP_850757.1 3 - 4 597 73
GenBank mRNA AB493733.1 - 2 - 3 337 73
AJ294532.1 - 2 - 3 337 73
AK228148.1 - 3 - 4 578 73

View Full Product Details