Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATPI4K ALPHA
Entrez Gene ID
841357
Gene Name
Phosphatidylinositol 3- and 4-kinase family protein
Gene Aliases
AT1G49340
Chromosome Location
Chr.1: 18252050 - 18264672 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103824.4 NP_175359.2 2 - 3 1647 107
NM_180629.2 NP_850960.1 2 - 3 1649 107

View Full Product Details