Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD12
Entrez Gene ID
830275
Gene Name
IQ-domain 12
Gene Aliases
AT5G03960, F8F6.170, F8F6_170, IQ-domain 12
Chromosome Location
Chr.5: 1068158 - 1070164 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120478.4 NP_196016.1 2 - 3 336 61

View Full Product Details