Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOL
Entrez Gene ID
830372
Gene Name
NYC1-like protein
Gene Aliases
AT5G04900, MUG13.25, MUG13_25, NYC1-like
Chromosome Location
Chr.5: 1434774 - 1437495 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120572.3 NP_568145.1 10 - 11 897 101
GenBank mRNA AB255027.1 - 10 - 11 845 101
AY085229.1 - 10 - 11 856 101
BT025238.1 - 10 - 11 845 101

View Full Product Details