Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHS
Entrez Gene ID
830477
Gene Name
deoxyhypusine synthase
Gene Aliases
AT5G05920, EDA22, K18J17.7, K18J17_7, deoxyhypusine synthase, embryo sac development arrest 22
Chromosome Location
Chr.5: 1777547 - 1779867 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342839.1 NP_001318486.1 2 - 3 804 103
NM_001342840.1 NP_001330195.1 3 - 4 649 103
NM_120674.4 NP_196211.1 3 - 4 754 103
GenBank mRNA AY087829.1 - 3 - 4 713 103

View Full Product Details