Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GUS2
Entrez Gene ID
830676
Gene Name
glucuronidase 2
Gene Aliases
AT5G07830, AtGUS2, F13G24.30, glucuronidase 2
Chromosome Location
Chr.5: 2503125 - 2506688 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120865.5 NP_196400.2 9 - 10 1649 98
GenBank mRNA BT015345.1 - 9 - 10 1430 98
BT015800.1 - 8 - 9 1250 98

View Full Product Details