Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BLH4
Entrez Gene ID
816908
Gene Name
BEL1-like homeodomain 4
Gene Aliases
AT2G23760, BEL1-like homeodomain 4, F27L4.6, F27L4_6, SAW2, SAWTOOTH 2
Chromosome Location
Chr.2: 10107707 - 10113327 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036327.2 NP_001031404.1 3 - 4 1514 70
NM_127939.3 NP_179956.1 2 - 3 1634 70
NM_179713.3 NP_850044.1 3 - 4 1726 70
GenBank mRNA AY058179.1 - 3 - 4 1530 70
AY142019.1 - 2 - 3 1352 70

View Full Product Details