Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCG37
- Entrez Gene ID
-
824516
- Gene Name
- pleiotropic drug resistance 9
- Gene Aliases
-
AT3G53480, ATP-binding cassette G37, ATPDR9, PDR9, PIS1, PLEIOTROPIC DRUG RESISTANCE 9, pleiotropic drug resistance 9, polar auxin transport inhibitor sensitive 1
- Chromosome Location
-
Chr.3: 19825267 - 19831880 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115208.4 |
NP_190916.1 |
15 - 16 |
2612 |
112 |
View Full Product Details