Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- U2AF35B
- Entrez Gene ID
-
834293
- Gene Name
- Zinc finger C-x8-C-x5-C-x3-H type family protein
- Gene Aliases
-
AT5G42820, ATU2AF35B, MJB21.20, MJB21_20
- Chromosome Location
-
Chr.5: 17169283 - 17172787 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_203141.2 |
NP_974870.1 |
2 - 3 |
1024 |
72 |
View Full Product Details