Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BZIP60
Entrez Gene ID
840897
Gene Name
basic region/leucine zipper motif 60
Gene Aliases
AT1G42990, ATBZIP60, Bbasic region/leucine zipper motif 60, F13A11.5, F13A11_5, basic region/leucine zipper motif 60
Chromosome Location
Chr.1: 16135723 - 16137463 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103458.3 NP_174998.1 1 - 2 445 67
GenBank mRNA AY045964.1 - 1 - 2 489 67
AY133805.1 - 1 - 2 441 67

View Full Product Details