Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHR18
Entrez Gene ID
841251
Gene Name
chromatin remodeling factor18
Gene Aliases
AT1G48310, CHA18, F11A17.14, F11A17_14, chromatin remodeling factor18
Chromosome Location
Chr.1: 17848421 - 17853915 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333322.1 NP_001321270.1 18 - 19 1648 75
NM_001333323.1 NP_001321271.1 18 - 19 1525 75
NM_001333324.1 NP_001321269.1 18 - 19 1627 75
NM_103728.5 NP_175265.3 18 - 19 1633 75

View Full Product Details