Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLA10
Entrez Gene ID
825261
Gene Name
FASCICLIN-like arabinogalactan-protein 10
Gene Aliases
AT3G60900, FASCICLIN-like arabinogalactan-protein 10
Chromosome Location
Chr.3: 22499292 - 22501239 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115954.3 NP_191649.1 1 - 1 1756 157
GenBank mRNA AY087634.1 - 1 - 1 1401 157

View Full Product Details