Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NDA2
- Entrez Gene ID
-
817549
- Gene Name
- alternative NAD(P)H dehydrogenase 2
- Gene Aliases
-
AT2G29990, F23F1.9, F23F1_9, alternative NAD(P)H dehydrogenase 2
- Chromosome Location
-
Chr.2: 12793320 - 12796085 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128553.3 |
NP_180560.1 |
4 - 5 |
1087 |
151 |
View Full Product Details