Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAC3
Entrez Gene ID
817571
Gene Name
laccase 3
Gene Aliases
AT2G30210, T9D9.2, laccase 3
Chromosome Location
Chr.2: 12887446 - 12889931 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128574.4 NP_180580.1 2 - 3 351 61

View Full Product Details