Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG40
Entrez Gene ID
831588
Gene Name
SET domain group 40
Gene Aliases
AT5G17240, MKP11.9, MKP11_9, SET domain group 40
Chromosome Location
Chr.5: 5666823 - 5668907 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343485.1 NP_001330251.1 3 - 4 680 75
NM_121730.4 NP_197226.2 4 - 5 601 75
GenBank mRNA AK175921.1 - 4 - 5 572 75
BT010455.1 - 4 - 5 570 75

View Full Product Details