Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KAPP
Entrez Gene ID
832048
Gene Name
kinase associated protein phosphatase
Gene Aliases
AT5G19280, RAG1, ROOT ATTENUATED GROWTH 1, T24G5.3, kinase associated protein phosphatase
Chromosome Location
Chr.5: 6488309 - 6493439 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161248.1 NP_001154720.1 1 - 2 272 104
NM_121933.4 NP_197429.1 1 - 2 282 104
GenBank mRNA AY045853.1 - 1 - 2 228 104
AY117152.1 - 1 - 2 141 104
U09505.1 - 1 - 2 179 104

View Full Product Details