Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UCC1
Entrez Gene ID
817791
Gene Name
uclacyanin 1
Gene Aliases
AT2G32300, uclacyanin 1
Chromosome Location
Chr.2: 13722362 - 13723641 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336386.1 NP_001325251.1 1 - 2 305 89
NM_128789.3 NP_180789.1 1 - 2 240 89
GenBank mRNA U76298.1 - 1 - 2 192 89

View Full Product Details