Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKT3
Entrez Gene ID
817876
Gene Name
peroxisomal 3-ketoacyl-CoA thiolase 3
Gene Aliases
3-KETOACYL-COA THIOLASE 2, AT2G33150, KAT2, PED1, PEROXISOME DEFECTIVE 1, peroxisomal 3-ketoacyl-CoA thiolase 3, potassium channel in Arabidopsis thaliana 2
Chromosome Location
Chr.2: 14047478 - 14051400 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128874.4 NP_180873.1 11 - 12 1501 92
GenBank mRNA AB008854.1 - 11 - 12 1192 92
AF327529.1 - 11 - 12 1192 92
AF349530.1 - 11 - 12 1084 92
AY052702.1 - 11 - 12 1212 92
AY063720.1 - 11 - 12 1084 92
AY087543.1 - 11 - 12 1160 92

View Full Product Details