Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APRR8
Entrez Gene ID
828011
Gene Name
pseudo-response regulator 8
Gene Aliases
AT4G00760, A_TM018A10.20, A_TM018A10_20, PRR8, PSEUDO-RESPONSE REGULATOR 8, T18A10.1, T18A10_1, pseudo-response regulator 8
Chromosome Location
Chr.4: 327025 - 333477 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340267.1 NP_001328422.1 1 - 2 345 78
NM_116302.4 NP_191985.3 9 - 10 2506 78
GenBank mRNA AK229225.1 - 9 - 10 2506 78

View Full Product Details