The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001340338.1 | NP_001328373.1 | 5 - 6 | 890 | 136 |
NM_116412.2 | NP_192091.1 | 5 - 6 | 901 | 136 |