Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APRL6
Entrez Gene ID
826471
Gene Name
APR-like 6
Gene Aliases
APR-like 6, AT4G08930, ATAPRL6, T3H13.4, T3H13_4
Chromosome Location
Chr.4: 5729169 - 5730850 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_116962.4 NP_192632.2 3 - 4 823 67

View Full Product Details