Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP1A
Entrez Gene ID
841846
Gene Name
oligouridylate-binding protein 1A
Gene Aliases
AT1G54080, F15I1.16, F15I1_16, oligouridylate-binding protein 1A
Chromosome Location
Chr.1: 20183545 - 20187096 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104285.5 NP_175810.1 4 - 5 641 90
NM_179475.1 NP_849806.1 4 - 5 454 90
GenBank mRNA AF348587.1 - 4 - 5 369 90
AF361639.1 - 4 - 5 515 90
AY133610.1 - 4 - 5 369 90

View Full Product Details