Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRX-LIKE3
Entrez Gene ID
841858
Gene Name
BREVIS RADIX-like 3
Gene Aliases
ARABIDOPSIS THALIANA BREVIS RADIX-LIKE 3, AT1G54180, ATBRXL3, BREVIS RADIX-like 3, F15I1.28, F15I1_28
Chromosome Location
Chr.1: 20227168 - 20230237 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160950.2 NP_001154422.1 2 - 3 526 73
NM_104296.5 NP_175820.2 2 - 3 403 73

View Full Product Details