Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ICDH
Entrez Gene ID
841875
Gene Name
isocitrate dehydrogenase
Gene Aliases
AT1G54340, F20D21.16, F20D21_16, isocitrate dehydrogenase
Chromosome Location
Chr.1: 20283326 - 20286786 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333641.1 NP_001321996.1 5 - 6 442 83
NM_104312.3 NP_175836.1 5 - 6 509 83
GenBank mRNA AF316501.1 - 5 - 6 335 83
BT025983.1 - 5 - 6 315 83

View Full Product Details