Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BCDH BETA1
- Entrez Gene ID
-
841998
- Gene Name
- branched-chain alpha-keto acid decarboxylase E1 beta subunit
- Gene Aliases
-
AT1G55510, BRANCHED-CHAIN ALPHA-KETO ACID DECARBOXYLASE E1 BETA SUBUNIT, T5A14.9, T5A14_9, branched-chain alpha-keto acid decarboxylase E1 beta subunit
- Chromosome Location
-
Chr.1: 20723279 - 20725747 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_104426.4 |
NP_175947.1 |
6 - 7 |
812 |
70 |
GenBank mRNA |
BT024741.1 |
- |
6 - 7 |
609 |
70 |
View Full Product Details